Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Sulfuric Acid Standard Solution, 0.020 N (N/50), 1 L

Catalog No. 501994202 Shop All Hach Company Products
Encompass

Sulfuric Acid Standard Solution, 0.020 N (N/50), 1 L

Catalog No. 50-199-4202 Supplier Hach Company Supplier No. 20353
May include imposed supplier surcharges.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Sulfuric Acid Standard Solution, 0.020 N (N/50), 1 L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.