Learn More
Hach Company Sulfuric Acid Standard Solution, 19.2 N, 500 mL
Supplier:  Hach Company 203849
Encompass
Multi-purpose sulfuric acid standard solution. Required for determination of volatile acids by the esterification method. 500 mL bottle
Catalog No. 50-199-4662