Learn More
Hach Company Sulfuric Acid Standard Solution, 5.25 N, 1 L
Supplier:  Hach Company 244953
Encompass
Multi-purpose sulfuric acid standard solution. Used for sample digestions and pH adjustments. 1 liter bottle
Catalog No. 50-199-4772