Learn More
Sulfuryl chloride, 1.0M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 378151000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Sulfuryl chloride | |
7791-25-5, 75-09-2 | |
Cl2O2S | |
MFCD00011451 | |
sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride | |
ClS(Cl)(=O)=O | |
134.96 | |
CHEBI:29291 | |
1M, exact strength on the certificate of analysis | |
Clear pale yellow solution | |
1.3520g/mL |
1.0M Solution in Dichloromethane | |
100 mL | |
SO2Cl2 | |
15, 9110 | |
YBBRCQOCSYXUOC-UHFFFAOYSA-N | |
sulfuryl dichloride | |
24648 | |
134.97 | |
AcroSeal™ Glass bottle | |
1.352 |
Safety and Handling
GHS H Statement
Suspected of causing cancer.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
Toxic if inhaled.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 232-245-6
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only