Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuryl chloride, 98.5%

Catalog No. AC169450010
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169450010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC169450010 Supplier Thermo Scientific Chemicals Supplier No. 169450010
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 7791-25-5 | Cl2O2S | 134.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-25-5
Molecular Formula Cl2O2S
Molecular Weight (g/mol) 134.96
MDL Number MFCD00011451
InChI Key YBBRCQOCSYXUOC-UHFFFAOYSA-N
Synonym sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride
PubChem CID 24648
ChEBI CHEBI:29291
IUPAC Name sulfuryl dichloride
SMILES ClS(Cl)(=O)=O

Specifications

Boiling Point 69.1°C
Melting Point -54.1°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 1 kg
Packaging Glass bottle
Refractive Index 1.443
Specific Gravity 1.667
Assay Percent Range 98% min. (ex Cl) (Argentometry)
Density 1.6670g/mL
Linear Formula SO2Cl2
Fieser 01,1128; 02,394; 03,276; 04,474; 05,641; 06,561; 10,375; 13,284; 14,291; 16,311
Merck Index 15,911
Solubility Information Solubility in water: reacts. Other solubilities: soluble in glacial acetic acid
Formula Weight 134.96
Percent Purity 98.5%
Chemical Name or Material Sulfuryl chloride, 98.5%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Fatal if inhaled.
Reacts violently with water.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED:
EINECSNumber 232-245-6
RTECSNumber WT4870000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.