Learn More
Apexbio Technology LLC Sulindac 38194-50-2 100mg
SDP
Catalog No. 5000093083
Encompass_Preferred
Sulindac (CAS 38194-50-2) is a nonsteroidal anti-inflammatory drug that functions as a prodrug and is metabolized in vivo to pharmacologically active sulindac sulfide and sulindac sulfone Sulindac sulfide acts as a cyclooxygenase inhibitor and modulates intracellular signaling pathways including the repression of ras activity Sulindac sulfone possesses antitumor properties independent of cyclooxygenase inhibition Due to these distinct metabolites and their activities sulindac is utilized in biomedical research for studying inflammatory processes cyclooxygenase-mediated pathways and the chemopreventive mechanisms involved in tumorigenesis
Catalog No. 50-000-93083 Supplier Apexbio Technology LLC Supplier No. B2044100
Sulindac (CAS 38194-50-2) is a nonsteroidal anti-inflammatory drug that functions as a prodrug and is metabolized in vivo to pharmacologically active sulindac sulfide and sulindac sulfone Sulindac sulfide acts as a cyclooxygenase inhibitor and modulates intracellular signaling pathways including the repression of ras activity Sulindac sulfone possesses antitumor properties independent of cyclooxygenase inhibition Due to these distinct metabolites and their activities sulindac is utilized in biomedical research for studying inflammatory processes cyclooxygenase-mediated pathways and the chemopreventive mechanisms involved in tumorigenesis