Learn More
Medchemexpress LLC Sulindac | 38194-50-2 | 99.68% | C20H17FO3S | 100 MG
SDP
Catalog No. 5000432255
Encompass_Preferred
Sulindac (MK-231) is an orally active nonsteroidal anti-inflammatory agent and an immunomodulatory agent. It is used for the research of arthritis of the spine, gouty arthritis, and various cancers, including colorectal cancer (CRC) and lung cancer.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.
Catalog No. 50-004-32255 Supplier Medchemexpress LLC Supplier No. HYB0008100MG
Sulindac (MK-231) is an orally active nonsteroidal anti-inflammatory agent and an immunomodulatory agent. It is used for the research of arthritis of the spine, gouty arthritis, and various cancers, including colorectal cancer (CRC) and lung cancer.
  • Store powder at -20°C for 3 years or 4°C for 2 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.