Learn More
Selleck Chemical LLC Sulindac-50mg
SDP
Catalog No. 502389774
Encompass_Preferred
Sulindac(MK-231) is a non-steroidal COX inhibitor, which potently inhibits prostaglandin synthesis, used in the treatment of acute or chronic inflammatory conditions.
Catalog No. 50-238-9774 Supplier Selleck Chemical LLC Supplier No. S200750MG
Sulindac(MK-231) is a non-steroidal COX inhibitor, which potently inhibits prostaglandin synthesis, used in the treatment of acute or chronic inflammatory conditions.