Learn More
Cayman Chemical SulIndac-d3 500ug
SDP
Catalog No. 502947958
Encompass_Preferred
An internal standard for the quantification of sulindac by GC- or LC-MS
Catalog No. 50-294-7958 Supplier Cayman Chemical Supplier No. 33381500 UG
An internal standard for the quantification of sulindac by GC- or LC-MS