Learn More
Cayman Chemical Sulindac Inhibitors
SDP
Supplier:  Cayman Chemical 100043861 G
Encompass_Preferred
An NSAID, a COX-2 inhibitor (IC50 = 0.79 µM), and a prodrug form of sulindac sulfide; selective for COX-2 over COX-1 (IC50 = >1,000 µM); inhibits acetic acid-induced writhing in mice and reduces carrageenan-induced paw edema in rats at 100 mg/kg; reduces tumor growth in a BxPC-3 mouse xenograft model at 60 mg/kg
Catalog No. 50-289-1104