Learn More
TARGETMOL CHEMICALS INC SULINDAC SULFIDE 10MG
SDP
Catalog No. 502581386
Encompass_Preferred
Also available in 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sulindac sulfide (cis-Sulindac sulfide) is a non-steroidal anti-inflammatory compound that has a high affinity for COX-1 and acts as an inhibitor in Ras activation of Raf-1. Sulindac sulfide is a non-competitive inhibitor of gamma-secretase with an IC50 value of 20.2 uM. It is an active metabolite of sulinic acid. purity: 99%
Catalog No. 50-258-1386 Supplier TARGETMOL CHEMICALS INC Supplier No. T1303710MG
Also available in 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sulindac sulfide (cis-Sulindac sulfide) is a non-steroidal anti-inflammatory compound that has a high affinity for COX-1 and acts as an inhibitor in Ras activation of Raf-1. Sulindac sulfide is a non-competitive inhibitor of gamma-secretase with an IC50 value of 20.2 uM. It is an active metabolite of sulinic acid. purity: 99%