Learn More
Cayman Chemical Sulindac sulfide, Purity: ≥98%, CAS: 49627-27-2, 5 mg
SDP
Catalog No. 502887653
Encompass_Preferred
An active metabolite of sulindac; inhibits COX-1 and COX-2 (IC50s = 1.9 and 1.21 µM, respectively); inhibits aldose reductase (IC50 = 279 nM), blocking NADPH-dependent reduction of glucose to sorbitol, and reducing type 2 diabetic complications; inhibits colorectal cancer growth in vitro and in vivo
Catalog No. 50-288-7653 Supplier Cayman Chemical Supplier No. 100043875 MG
An active metabolite of sulindac; inhibits COX-1 and COX-2 (IC50s = 1.9 and 1.21 µM, respectively); inhibits aldose reductase (IC50 = 279 nM), blocking NADPH-dependent reduction of glucose to sorbitol, and reducing type 2 diabetic complications; inhibits colorectal cancer growth in vitro and in vivo