Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sulindac sulfide | 49627-27-2 | 98.1% | 100 MG
SDP

Catalog No. 5000412015
Encompass_Preferred

Sulindac sulfide is a noncompetitive γ-secretase inhibitor, with an IC50 of 20.2 μM for γ42-secretase activity. For research use only. MedChemExpress does not sell to patients.

  • Solubility in vitro: DMSO at 50 mg/mL (146.88 mM), requires ultrasonic assistance, use newly opened DMSO due to hygroscopic nature.
  • Stock solution storage: -80°C for 6 months; -20°C for 1 month.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage of powder: -20°C for 3 years; in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-004-12015 Supplier Medchemexpress LLC Supplier No. HYB1786100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sulindac sulfide is a noncompetitive γ-secretase inhibitor, with an IC50 of 20.2 μM for γ42-secretase activity. For research use only. MedChemExpress does not sell to patients.

  • Solubility in vitro: DMSO at 50 mg/mL (146.88 mM), requires ultrasonic assistance, use newly opened DMSO due to hygroscopic nature.
  • Stock solution storage: -80°C for 6 months; -20°C for 1 month.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage of powder: -20°C for 3 years; in solvent: -80°C for 6 months, -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.