Learn More
Medchemexpress LLC Sulindac sulfide | 49627-27-2 | 98.10% | 250 MG
SDP
Catalog No. 5000414414
Encompass_Preferred
Sulindac sulfide is a noncompetitive γ-secretase inhibitor, demonstrating an IC50 of 20.2 μM for γ42-secretase activity. This compound is primarily used in research, especially for studying neurological diseases.
  • Noncompetitive γ-secretase inhibitor
  • IC50 of 20.2 μM for γ42-secretase activity
  • Suitable for research applications
  • Useful for studying neurological diseases
Catalog No. 50-004-14414 Supplier Medchemexpress LLC Supplier No. 250MGHYB1786
Sulindac sulfide is a noncompetitive γ-secretase inhibitor, demonstrating an IC50 of 20.2 μM for γ42-secretase activity. This compound is primarily used in research, especially for studying neurological diseases.
  • Noncompetitive γ-secretase inhibitor
  • IC50 of 20.2 μM for γ42-secretase activity
  • Suitable for research applications
  • Useful for studying neurological diseases