Learn More
TARGETMOL CHEMICALS INC Sulindac sulfone 25MG
SDP
Catalog No. 502879190
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.Sulindac sulfone is a metabolite of the nonsteroidal anti-inflammatory drug sulindac. Sulindac sulfone is an inhibitor of aldose reductase (IC50 =367 nM). Purity 98.19%
Catalog No. 50-287-9190 Supplier TARGETMOL CHEMICALS INC Supplier No. T2340325MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes.Sulindac sulfone is a metabolite of the nonsteroidal anti-inflammatory drug sulindac. Sulindac sulfone is an inhibitor of aldose reductase (IC50 =367 nM). Purity 98.19%