Learn More
Enzo Life Sciences Sulindac sulfone (25mg). CAS: 59864-04-9
SDP
Catalog No. 502010312
Encompass_Preferred
Induces apoptosis but does not inhibit cyclooxygenase. Inhibits growth and induces apoptosis in human cancer cell lines. Metabolite of Sulindac (ALX-430-105). Alternative name: Z-5-Fluoro-2-methyl-1-[p-(methyl-sulfonyl)benzylidene]indene-3-acetic acid. Purity: ≥94% (HPLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (3mg/ml, warm). Long Term Storage: Ambient.
Catalog No. 50-201-0312 Supplier Enzo Life Sciences Supplier No. BMLAP2010025
Induces apoptosis but does not inhibit cyclooxygenase. Inhibits growth and induces apoptosis in human cancer cell lines. Metabolite of Sulindac (ALX-430-105). Alternative name: Z-5-Fluoro-2-methyl-1-[p-(methyl-sulfonyl)benzylidene]indene-3-acetic acid. Purity: ≥94% (HPLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (3mg/ml, warm). Long Term Storage: Ambient.