Learn More
Apexbio Technology LLC Sulindac sulfone 59864-04-9 5mg
SDP
Catalog No. 5000091318
Encompass_Preferred
Sulindac sulfone (CAS 59864-04-9) is a metabolite of the NSAID sulindac and has been investigated for its effects on colorectal cancer models It modulates cellular signaling by decreasing -catenin pro-caspase 3 cyclin D1 and PPAR levels in SW480 cells and reduces TCF-mediated transcription in a dose-dependent manner In vitro sulindac sulfone inhibits PGE2 production in HCA-7 cells (IC50 360 mmol/L) and decreases colony formation in HCA-7 and HCT-116 cells (EC50 50 mmol/L) In vivo it lowers tumor incidence in the AOM-induced colorectal cancer rat model Clinically it has led to regression of rectal polyps in familial adenomatous polyposis patients Sulindac sulfone is utilized in research focused on colorectal cancer chemoprevention and -catenin pathway modulation
Catalog No. 50-000-91318 Supplier Apexbio Technology LLC Supplier No. C34535
Sulindac sulfone (CAS 59864-04-9) is a metabolite of the NSAID sulindac and has been investigated for its effects on colorectal cancer models It modulates cellular signaling by decreasing -catenin pro-caspase 3 cyclin D1 and PPAR levels in SW480 cells and reduces TCF-mediated transcription in a dose-dependent manner In vitro sulindac sulfone inhibits PGE2 production in HCA-7 cells (IC50 360 mmol/L) and decreases colony formation in HCA-7 and HCT-116 cells (EC50 50 mmol/L) In vivo it lowers tumor incidence in the AOM-induced colorectal cancer rat model Clinically it has led to regression of rectal polyps in familial adenomatous polyposis patients Sulindac sulfone is utilized in research focused on colorectal cancer chemoprevention and -catenin pathway modulation