Learn More
Medchemexpress LLC Sulindac sulfone | 59973-80-7 | 98.1% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB17875MG
Encompass_Preferred
Sulindac sulfone is an mTORC1 pathway inhibitor and a metabolite of Sulindac. It inhibits colon cancer cell growth and induces cell cycle arrest. Sulindac sulfone is utilized in cancer research.
  • Cytotoxicity against human COLO320 cells after 72 hrs by WST-1 assay (IC50 > 200 μM)
  • Cytotoxicity against human MIAPaCa2 cells after 72 hrs by WST-1 assay (IC50 > 200 μM)
Catalog No. 50-004-12863