Learn More
Enzo Life Sciences Sulindac sulfone (5mg). CAS: 59864-04-9
SDP
Catalog No. 502010311
Encompass_Preferred
Induces apoptosis but does not inhibit cyclooxygenase. Inhibits growth and induces apoptosis in human cancer cell lines. Metabolite of Sulindac (ALX-430-105). Alternative name: Z-5-Fluoro-2-methyl-1-[p-(methyl-sulfonyl)benzylidene]indene-3-acetic acid. Purity: ≥94% (HPLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (3mg/ml, warm). Long Term Storage: Ambient.
Catalog No. 50-201-0311 Supplier Enzo Life Sciences Supplier No. BMLAP2010005
Add to Cart
Add to Cart
Induces apoptosis but does not inhibit cyclooxygenase. Inhibits growth and induces apoptosis in human cancer cell lines. Metabolite of Sulindac (ALX-430-105). Alternative name: Z-5-Fluoro-2-methyl-1-[p-(methyl-sulfonyl)benzylidene]indene-3-acetic acid. Purity: ≥94% (HPLC). Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (3mg/ml, warm). Long Term Storage: Ambient.