Learn More
Cayman Chemical Sulndac-d3 1mg
SDP
Catalog No. 502950528
Encompass_Preferred
An internal standard for the quantification of sulindac by GC- or LC-MS
Catalog No. 50-295-0528 Supplier Cayman Chemical Supplier No. 333811 MG
An internal standard for the quantification of sulindac by GC- or LC-MS