Learn More
Selleck Chemical LLC Suloctidil-25mg
SDP
Supplier:  Selleck Chemical LLC S527125MG
Encompass_Preferred
Suloctidil, a sulfur-containing aminoalcohol showing to inhibit human platelet aggregation, is a peripheral vasodilator and a potential antifungal agent.
Catalog No. 50-239-0038