Learn More
Lovibond™ Sulphite Drop Test Kit
Catalog No. 09051499 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
20 mL
Catalog No. Volume (Metric)
09-051-499 20 mL
1 options
Catalog No. 09-051-499 Supplier Lovibond™ Supplier No. 56K700360
Add to Cart
Add to Cart
Comes housed in its own rugged carry case, complete with all necessary equipment such as instructions, reagents, syringe and test bottle or test cell for 100 tests.
  • Ideal for mobile treatment engineers
  • Compact and portable design
  • Easy to follow instructions including pictograms
  • Complete with all necessary equipment
Specifications
pH Range 0 to 14
Volume (Metric) 20 mL
No. of Tests 100 Tests
Type Sulphite Test