Learn More
Cayman Chemical Sulpho Nonate 10mg
SDP
Catalog No. 502896411
Encompass_Preferred
Sulpho NONOate produces nitrous oxide but no NO at physiological pH. Therefore, this compound may be used as a negative control in experiments using other NO donors of the NONOate class.{875,5437}
Catalog No. 50-289-6411 Supplier Cayman Chemical Supplier No. 8330010 MG
Sulpho NONOate produces nitrous oxide but no NO at physiological pH. Therefore, this compound may be used as a negative control in experiments using other NO donors of the NONOate class.{875,5437}