Learn More
Cayman Chemical Sulpho NONOate, Purity: ≥90%, CAS: 61142-90-3, 50 mg
SDP
Catalog No. 502896596
Encompass_Preferred
Sulpho NONOate produces nitrous oxide but no NO at physiological pH. Therefore, this compound may be used as a negative control in experiments using other NO donors of the NONOate class.{875,5437}
Catalog No. 50-289-6596 Supplier Cayman Chemical Supplier No. 8330050 MG
Sulpho NONOate produces nitrous oxide but no NO at physiological pH. Therefore, this compound may be used as a negative control in experiments using other NO donors of the NONOate class.{875,5437}