Learn More
Aviva Systems Biology Corp SULT1A1 Antibody - N-terminal region (ARP49134_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49134_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SULT1A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SULT1A1. Alternate names: HAST1/HAST2, MGC131921, MGC5163, P-PST, PST, ST1A3, STP, STP1, TSPST1, ST1A1ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60029