Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SULT1A1 Human His 250ug
SDP

Catalog No. 5000197023
Encompass_Preferred

SULT1A1 Protein, Human (His) is a recombinant, full-length human sulfotransferase produced in Escherichia coli with an N-terminal 6xHis affinity tag. Supplied as a stabilized, 0.2 μm filtered solution (250 μg), it is designed for biochemical and enzymatic assays and is characterized by high purity and measured enzymatic activity.

  • Full-length human SULT1A1 expressed in E. coli.
  • N-terminal 6xHis tag for affinity purification.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Specific activity 56.4 pmol/min/μg (PAPS/1-naphthol assay).
  • Stabilized formulation with trehalose, glycerol, and Tween 80 for storage.
  • Store at -80°C; avoid repeated freeze-thaw cycles.

Catalog No. 50-001-97023 Supplier Medchemexpress LLC Supplier No. HYP70969250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SULT1A1 Protein, Human (His) is a recombinant, full-length human sulfotransferase produced in Escherichia coli with an N-terminal 6xHis affinity tag. Supplied as a stabilized, 0.2 μm filtered solution (250 μg), it is designed for biochemical and enzymatic assays and is characterized by high purity and measured enzymatic activity.

  • Full-length human SULT1A1 expressed in E. coli.
  • N-terminal 6xHis tag for affinity purification.
  • High purity (>95%) as determined by reducing SDS-PAGE.
  • Specific activity 56.4 pmol/min/μg (PAPS/1-naphthol assay).
  • Stabilized formulation with trehalose, glycerol, and Tween 80 for storage.
  • Store at -80°C; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.