Learn More
ABclonal Technology SULT1A1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15991000UL
Encompass_Preferred
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes one of two phenol sulfotransferases with thermostable enzyme activity. Multiple alternatively spliced variants that encode two isoforms have been identified for this gene.
Catalog No. 50-001-04584