Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SULT1A3 Human N-Hi 1mg
SDP

Catalog No. 5000196533
Encompass_Preferred

Recombinant human SULT1A3 is a sulfotransferase enzyme supplied as an N-terminal His-tagged protein produced in E. coli and provided as a lyophilized powder for biochemical and enzymatic studies. The enzyme catalyzes sulfate conjugation of phenolic monoamines (for example dopamine and serotonin) and select xenobiotics, making it suitable for metabolism, drug interaction, and neurotransmitter regulation assays.

  • Human sequence, M1-L295, representing endogenous enzyme.
  • Expressed in E. coli with an N-terminal 6xHis tag for affinity purification.
  • Lyophilized powder suitable for long-term storage.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight ~35 kDa.
  • Recommend reconstitution to ≥100 μg/mL in ddH2O.
  • Available in multiple package sizes including 1 mg.

Catalog No. 50-001-96533 Supplier Medchemexpress LLC Supplier No. HYP71341A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human SULT1A3 is a sulfotransferase enzyme supplied as an N-terminal His-tagged protein produced in E. coli and provided as a lyophilized powder for biochemical and enzymatic studies. The enzyme catalyzes sulfate conjugation of phenolic monoamines (for example dopamine and serotonin) and select xenobiotics, making it suitable for metabolism, drug interaction, and neurotransmitter regulation assays.

  • Human sequence, M1-L295, representing endogenous enzyme.
  • Expressed in E. coli with an N-terminal 6xHis tag for affinity purification.
  • Lyophilized powder suitable for long-term storage.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight ~35 kDa.
  • Recommend reconstitution to ≥100 μg/mL in ddH2O.
  • Available in multiple package sizes including 1 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.