Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SULT1A3 Human N-Hi 50ug
SDP

Catalog No. 5000196894
Encompass_Preferred

Recombinant human SULT1A3 protein with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied lyophilized for biochemical and enzymatic studies. The enzyme catalyzes PAPS-dependent sulfate conjugation of phenolic monoamines (for example dopamine, norepinephrine, and serotonin) and is provided with high purity and a stabilized formulation for storage and handling.

  • Recombinant human sequence, full-length isoform 1.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Expressed in E. coli for efficient production.
  • Lyophilized formulation for improved shelf stability.
  • Formulated in PBS with trehalose, mannitol, and Tween 80 for protection during storage.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Supplied in microgram quantities suitable for biochemical assays.
  • Recommended storage at -20 °C to -80 °C; aliquot after reconstitution.

Catalog No. 50-001-96894 Supplier Medchemexpress LLC Supplier No. HYP71341A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human SULT1A3 protein with an N-terminal 6xHis tag, expressed in Escherichia coli and supplied lyophilized for biochemical and enzymatic studies. The enzyme catalyzes PAPS-dependent sulfate conjugation of phenolic monoamines (for example dopamine, norepinephrine, and serotonin) and is provided with high purity and a stabilized formulation for storage and handling.

  • Recombinant human sequence, full-length isoform 1.
  • N-terminal 6xHis tag for affinity purification and detection.
  • Expressed in E. coli for efficient production.
  • Lyophilized formulation for improved shelf stability.
  • Formulated in PBS with trehalose, mannitol, and Tween 80 for protection during storage.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Supplied in microgram quantities suitable for biochemical assays.
  • Recommended storage at -20 °C to -80 °C; aliquot after reconstitution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.