Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SULT1A3 Human N-Hi 5ug
SDP

Catalog No. 5000196756
Encompass_Preferred

Recombinant human SULT1A3 is a PAPS-dependent sulfotransferase expressed in E. coli and supplied as an N-terminal His-tagged, lyophilized protein. The 295-amino-acid protein (~35 kDa) is >95% pure and has measured activity in 1-naphthol sulfation assays, making it suitable for biochemical characterization and enzymatic studies.

  • Expressed in E. coli with N-terminal His tag.
  • 295 amino acids, approximately 35 kDa.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation with PBS, trehalose, mannitol, and Tween 80.
  • Specific activity reported for PAPS-dependent sulfation assays.
  • Stable at -20°C or -80°C for extended storage; aliquot after reconstitution.
  • Supplied as a 5 μg unit suitable for small-scale assays.
  • Applicable to enzymatic assays, inhibitor screening, and antibody development.

Catalog No. 50-001-96756 Supplier Medchemexpress LLC Supplier No. HYP71341A5UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human SULT1A3 is a PAPS-dependent sulfotransferase expressed in E. coli and supplied as an N-terminal His-tagged, lyophilized protein. The 295-amino-acid protein (~35 kDa) is >95% pure and has measured activity in 1-naphthol sulfation assays, making it suitable for biochemical characterization and enzymatic studies.

  • Expressed in E. coli with N-terminal His tag.
  • 295 amino acids, approximately 35 kDa.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation with PBS, trehalose, mannitol, and Tween 80.
  • Specific activity reported for PAPS-dependent sulfation assays.
  • Stable at -20°C or -80°C for extended storage; aliquot after reconstitution.
  • Supplied as a 5 μg unit suitable for small-scale assays.
  • Applicable to enzymatic assays, inhibitor screening, and antibody development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.