Learn More
Aviva Systems Biology Corp SULT1C2 Antibody - C-terminal region (ARP49130_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49130_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SULT1C2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SULT1C2. Alternate names: ST1C1, ST1C2, SULT1C1, humSULTC2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60027