Learn More
Aviva Systems Biology Corp SULT1C4 Antibody - middle region (ARP48707_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48707_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SULT1C4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SULT1C4. Alternate names: MGC149521, MGC34422, SULT1C, SULT1C2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit
Catalog No. 50-014-59893