Learn More
Aviva Systems Biology Corp SULT1E1 Antibody - middle region (ARP48669_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48669_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SULT1E1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SULT1E1. Alternate names: EST, EST-1, MGC34459, STE, ST1E1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-374