Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SULT1E1 protein, human (His) | >95.0% | 50 UG
SDP

Catalog No. 5000290943
Encompass_Preferred

Recombinant human SULT1E1 (estrogen sulfotransferase) with an N-terminal His tag, expressed in E. coli and purified to >95% for biochemical and functional studies. Supplied in multiple sizes, including a 50 μg vial; refer to the datasheet and COA for batch-specific concentration and storage instructions.

  • Recombinant human enzyme suitable for activity and structural assays.
  • N-terminal His tag enables straightforward purification and detection.
  • Expressed in E. coli for scalable, reproducible production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Available in small-volume sizes for low-consumption experiments.
  • Stable when stored at -20°C; aliquot and store at -80°C for extended stability.
  • Provided with batch-specific certificate of analysis for traceability.

Catalog No. 50-002-90943 Supplier Medchemexpress LLC Supplier No. HYP76099A50UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human SULT1E1 (estrogen sulfotransferase) with an N-terminal His tag, expressed in E. coli and purified to >95% for biochemical and functional studies. Supplied in multiple sizes, including a 50 μg vial; refer to the datasheet and COA for batch-specific concentration and storage instructions.

  • Recombinant human enzyme suitable for activity and structural assays.
  • N-terminal His tag enables straightforward purification and detection.
  • Expressed in E. coli for scalable, reproducible production.
  • Purity greater than 95% as determined by reducing SDS-PAGE.
  • Available in small-volume sizes for low-consumption experiments.
  • Stable when stored at -20°C; aliquot and store at -80°C for extended stability.
  • Provided with batch-specific certificate of analysis for traceability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.