Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SULT1E1 protein, human (His) | >95.0% | 1 MG
SDP

Catalog No. 5000291931
Encompass_Preferred

Recombinant human sulfotransferase SULT1E1 with an N-terminal 6xHis affinity tag, provided lyophilized for biochemical and enzymatic research applications such as estrogen metabolism and sulfation assays. Expressed in E. coli and purified to >95% purity, the protein is formulated for stability and intended for research use only.

  • Recombinant human enzyme with N-terminal 6xHis tag.
  • Expressed in E. coli for high-yield production.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation in 50 mM Tris-HCl, 200 mM NaCl, pH 7.4, 8% trehalose.
  • Stable at -20°C; aliquot and store at -20°C or -80°C after reconstitution.
  • Suitable for biochemical assays and estrogen sulfation studies.

Catalog No. 50-002-91931 Supplier Medchemexpress LLC Supplier No. HYP76099A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human sulfotransferase SULT1E1 with an N-terminal 6xHis affinity tag, provided lyophilized for biochemical and enzymatic research applications such as estrogen metabolism and sulfation assays. Expressed in E. coli and purified to >95% purity, the protein is formulated for stability and intended for research use only.

  • Recombinant human enzyme with N-terminal 6xHis tag.
  • Expressed in E. coli for high-yield production.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Lyophilized formulation in 50 mM Tris-HCl, 200 mM NaCl, pH 7.4, 8% trehalose.
  • Stable at -20°C; aliquot and store at -20°C or -80°C after reconstitution.
  • Suitable for biochemical assays and estrogen sulfation studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.