Learn More
Medchemexpress LLC SULT1E1 protein, human (N-His) | >95.0% | 500 UG
SDP
Catalog No. 5000291930
Encompass_Preferred
Recombinant human SULT1E1 (N-His) is a purified sulfotransferase enzyme expressed in Escherichia coli and supplied as a sterile, lyophilized powder. The protein is characterized by >95% purity by reducing SDS-PAGE, is formulated from 50 mM Tris-HCl, 200 mM NaCl, pH 7.4 with 8% trehalose, and is provided in sizes suitable for biochemical assays and enzyme characterization.
  • Recombinant human enzyme with N-terminal His tag
  • Expressed in Escherichia coli for consistent expression yield
  • Greater than 95.0% purity by reducing SDS-PAGE
  • Lyophilized sterile powder for long-term storage
  • Lyophilized from 50 mM Tris-HCl, 200 mM NaCl, pH 7.4, 8% trehalose
  • Recommended reconstitution concentration not less than 100 μg/mL in ddH2O
Catalog No. 50-002-91930 Supplier Medchemexpress LLC Supplier No. HYP76099A500UG
Recombinant human SULT1E1 (N-His) is a purified sulfotransferase enzyme expressed in Escherichia coli and supplied as a sterile, lyophilized powder. The protein is characterized by >95% purity by reducing SDS-PAGE, is formulated from 50 mM Tris-HCl, 200 mM NaCl, pH 7.4 with 8% trehalose, and is provided in sizes suitable for biochemical assays and enzyme characterization.
  • Recombinant human enzyme with N-terminal His tag
  • Expressed in Escherichia coli for consistent expression yield
  • Greater than 95.0% purity by reducing SDS-PAGE
  • Lyophilized sterile powder for long-term storage
  • Lyophilized from 50 mM Tris-HCl, 200 mM NaCl, pH 7.4, 8% trehalose
  • Recommended reconstitution concentration not less than 100 μg/mL in ddH2O