Learn More
Sino Biological SULT2A1 Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 11411R004100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human SULT2A1 (rh SULT2A1; Catalog#11411-H07E; NP_003158.2; Ser 2-Glu 285).
Catalog No. 50-224-7874