Learn More
Sino Biological SULT2A1 Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11411RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human SULT2A1 (rh SULT2A1; Catalog#11411-H07E; NP_003158.2; Ser 2-Glu 285). SULT2A1 specific IgG was purified by Human SULT2A1 affinity chromatography.
Catalog No. 50-224-8625