Learn More
Sino Biological SULT2A1 / STD Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11411MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SULT2A1 (rh SULT2A1; Catalog#11411-H07E; NP_003158.2; Ser 2-Glu 285). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8622