Learn More
Aviva Systems Biology Corp SULT2B1 Antibody - N-terminal region (ARP51155_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP51155_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SULT2B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SULT2B1. Alternate names: HSST2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60501