Learn More
Aviva Systems Biology Corp SULT2B1 Antibody - middle region (ARP48635_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48635_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SULT2B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SULT2B1. Alternate names: HSST2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59874