Learn More
Aviva Systems Biology Corp SULT2B1 Antibody - C-terminal region (ARP48636_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48636_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SULT2B1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SULT2B1. Alternate names: HSST2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59875