Learn More
Medchemexpress LLC Sultamicillin | 76497-13-7 | 50 MG
SDP
Catalog No. 5000417651
Encompass_Preferred
Sultamicillin is a broad-spectrum, orally active beta-lactamase inhibitor and an antibiotic with antibacterial activity. It is for research use only and exhibits antibacterial activity even against MRSA.
  • Broad-spectrum beta-lactamase inhibitor
  • Orally active antibiotic
  • Exhibits antibacterial activity, including against MRSA
  • Hydrolyzed to Sulbactam and Ampicillin upon oral absorption
  • Can improve survival in experimental polymicrobial sepsis in mice when combined with immunosuppression agents
  • Enhances bacterial clearance
Catalog No. 50-004-17651 Supplier Medchemexpress LLC Supplier No. HYN711550MG
Sultamicillin is a broad-spectrum, orally active beta-lactamase inhibitor and an antibiotic with antibacterial activity. It is for research use only and exhibits antibacterial activity even against MRSA.
  • Broad-spectrum beta-lactamase inhibitor
  • Orally active antibiotic
  • Exhibits antibacterial activity, including against MRSA
  • Hydrolyzed to Sulbactam and Ampicillin upon oral absorption
  • Can improve survival in experimental polymicrobial sepsis in mice when combined with immunosuppression agents
  • Enhances bacterial clearance