Learn More
Medchemexpress LLC Sultamicillin | 76497-13-7 | MFCD00864948 | 100 MG
SDP
Catalog No. 5000419130
Encompass_Preferred
Sultamicillin is a broad-spectrum and orally active beta-lactamase inhibitor, functioning as an antibiotic with antibacterial activity.
  • Broad-spectrum and orally active beta-lactamase inhibitor
  • Exhibits antibacterial activity
  • Hydrolyzed to sulbactam and ampicillin upon oral absorption
  • Effective against MRSA with a MIC value of 25 μg/mL
  • Improves survival and enhances bacterial clearance in experimental polymicrobial sepsis when combined with immunosuppression agents
Catalog No. 50-004-19130 Supplier Medchemexpress LLC Supplier No. HYN7115100MG
Sultamicillin is a broad-spectrum and orally active beta-lactamase inhibitor, functioning as an antibiotic with antibacterial activity.
  • Broad-spectrum and orally active beta-lactamase inhibitor
  • Exhibits antibacterial activity
  • Hydrolyzed to sulbactam and ampicillin upon oral absorption
  • Effective against MRSA with a MIC value of 25 μg/mL
  • Improves survival and enhances bacterial clearance in experimental polymicrobial sepsis when combined with immunosuppression agents