Learn More
Selleck Chemical LLC Sultamicillin Tosylate S5406-25mg
SDP
Catalog No. 503108562
Encompass_Preferred
Sultamicillin Tosylate is the tosylate salt of the double ester of sulbactam plus ampicillin Sulbactam is a semisynthetic beta-lactamase inhibitor which in combination with ampicillin extends the antibacterial activity of the latter to include some beta-lactamase-producing strains of bacteria that would otherwise be resistant
Catalog No. 50-310-8562 Supplier Selleck Chemical LLC Supplier No. S540625MG
Sultamicillin Tosylate is the tosylate salt of the double ester of sulbactam plus ampicillin Sulbactam is a semisynthetic beta-lactamase inhibitor which in combination with ampicillin extends the antibacterial activity of the latter to include some beta-lactamase-producing strains of bacteria that would otherwise be resistant