Learn More
Medchemexpress LLC Sultamicillin tosylate | 83105-70-8 | 1 ML
SDP
Catalog No. 5000418292
Encompass_Preferred
Sultamicillin tosylate is a broad-spectrum and orally active beta-lactamase inhibitor, and an antibiotic with antibacterial activity. It is for research use only.
  • Acts as a broad-spectrum and orally active beta-lactamase inhibitor.
  • Exhibits antibacterial activity.
  • Shows activity against MRSA with a MIC value of 25 μg/mL in vitro.
  • Hydrolyzes to Sulbactam and Ampicillin upon oral absorption.
  • Improves survival in experimental polymicrobial sepsis models in mice.
  • Enhances bacterial clearance when combined with immunosuppression agents.
  • Storage (solid): 4°C, sealed, away from moisture and light.
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month, sealed, away from moisture and light.
Catalog No. 50-004-18292 Supplier Medchemexpress LLC Supplier No. HYN711110MM/1ML
Sultamicillin tosylate is a broad-spectrum and orally active beta-lactamase inhibitor, and an antibiotic with antibacterial activity. It is for research use only.
  • Acts as a broad-spectrum and orally active beta-lactamase inhibitor.
  • Exhibits antibacterial activity.
  • Shows activity against MRSA with a MIC value of 25 μg/mL in vitro.
  • Hydrolyzes to Sulbactam and Ampicillin upon oral absorption.
  • Improves survival in experimental polymicrobial sepsis models in mice.
  • Enhances bacterial clearance when combined with immunosuppression agents.
  • Storage (solid): 4°C, sealed, away from moisture and light.
  • Storage (in solvent): -80°C for 6 months; -20°C for 1 month, sealed, away from moisture and light.