Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sultiame | 61-56-3 | 98.9% | 290.36 g/mol | C10H14N2O4S2 | 100 MG
SDP

Catalog No. 5000243680
Encompass_Preferred

Sultiame is a carbonic anhydrase inhibitor widely used as an antiepileptic agent. This product is provided as research-grade analytical material intended for laboratory studies, assay calibration, and pharmacology investigations.

  • Provides carbonic anhydrase inhibition for mechanism-of-action studies.
  • Used as an antiepileptic agent in pharmacology and toxicology research.
  • Analytical standard suitable for assay, calibration, and reference use.
  • High purity, approximately 98.9%.
  • Molecular weight 290.36 g/mol; formula C10H14N2O4S2.
  • CAS number 61-56-3 for unambiguous identification.
  • Available as solid and as a 10 mM DMSO solution for ready-to-use preparations.
  • Multiple pack sizes available, including 100 MG solid.

Catalog No. 50-002-43680 Supplier Medchemexpress LLC Supplier No. HY108316100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sultiame is a carbonic anhydrase inhibitor widely used as an antiepileptic agent. This product is provided as research-grade analytical material intended for laboratory studies, assay calibration, and pharmacology investigations.

  • Provides carbonic anhydrase inhibition for mechanism-of-action studies.
  • Used as an antiepileptic agent in pharmacology and toxicology research.
  • Analytical standard suitable for assay, calibration, and reference use.
  • High purity, approximately 98.9%.
  • Molecular weight 290.36 g/mol; formula C10H14N2O4S2.
  • CAS number 61-56-3 for unambiguous identification.
  • Available as solid and as a 10 mM DMSO solution for ready-to-use preparations.
  • Multiple pack sizes available, including 100 MG solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.