Learn More
Medchemexpress LLC Sultiame | 61-56-3 | MFCD00865304 | 290.36 | C10H14N2O4S2 | 50 MG
SDP
Catalog No. 5000244301
Encompass_Preferred
Sultiame is a sulfonamide carbonic anhydrase inhibitor used as an antiepileptic agent and supplied as an analytical standard for laboratory research. It is intended for use in method development, qualitative and quantitative analyses, and other research applications.
  • Carbonic anhydrase inhibitor with antiepileptic activity.
  • Provided as an analytical standard for research and method development.
  • Suitable for HPLC, GC, and MS qualitative and quantitative analyses.
  • Supplied as a solid compatible with common laboratory preparation methods.
  • Molecular formula C10H14N2O4S2; molecular weight 290.36.
  • Available in small scales appropriate for analytical use (for example, 5-500 mg).
Catalog No. 50-002-44301 Supplier Medchemexpress LLC Supplier No. HY10831650MG
Sultiame is a sulfonamide carbonic anhydrase inhibitor used as an antiepileptic agent and supplied as an analytical standard for laboratory research. It is intended for use in method development, qualitative and quantitative analyses, and other research applications.
  • Carbonic anhydrase inhibitor with antiepileptic activity.
  • Provided as an analytical standard for research and method development.
  • Suitable for HPLC, GC, and MS qualitative and quantitative analyses.
  • Supplied as a solid compatible with common laboratory preparation methods.
  • Molecular formula C10H14N2O4S2; molecular weight 290.36.
  • Available in small scales appropriate for analytical use (for example, 5-500 mg).