Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sultopride hydrochloride | 23694-17-9 | 99.9% | 390.93 | 1 ML
SDP

Catalog No. 5000403580
Encompass_Preferred

Sultopride hydrochloride is a selective antagonist of dopamine D2 receptor.

  • Selective antagonist of dopamine D2 receptor.
  • Purity of 99.92%.
  • Molecular weight of 390.93.
  • Appears as a white to off-white solid.
  • Soluble in H2O and DMSO.

Catalog No. 50-004-03580 Supplier Medchemexpress LLC Supplier No. HY42849A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sultopride hydrochloride is a selective antagonist of dopamine D2 receptor.

  • Selective antagonist of dopamine D2 receptor.
  • Purity of 99.92%.
  • Molecular weight of 390.93.
  • Appears as a white to off-white solid.
  • Soluble in H2O and DMSO.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.