Learn More
Medchemexpress LLC Sultopride hydrochloride | 23694-17-9 | 99.9% | 390.93 | 25 MG
SDP
Catalog No. 5000403414
Encompass_Preferred
Sultopride hydrochloride is a selective antagonist of the dopamine D2 receptor.
  • Selective antagonist of dopamine D2 receptor.
  • Has shown higher levels of DOPAC and HVA in the striatum and nucleus accumbens in rats compared to sulpiride-treated rats.
  • Available for research use only.
Catalog No. 50-004-03414 Supplier Medchemexpress LLC Supplier No. 25MGHY42849A
Sultopride hydrochloride is a selective antagonist of the dopamine D2 receptor.
  • Selective antagonist of dopamine D2 receptor.
  • Has shown higher levels of DOPAC and HVA in the striatum and nucleus accumbens in rats compared to sulpiride-treated rats.
  • Available for research use only.